TBS with Tween® (TBST), 20X solution, Molecular Biology Grade, ultrapure
Supplier: Thermo Fisher Scientific
Certificates
About this item
TBS is used in Western blotting protocols.
- Contaminant Testing:
- DNase (endo): Passes test
- DNase (exo): Passes test
- Protease: Passes test
Solution is 0,2 μm filtered and is supplied in an upright carton with a convenient dispensing spout.
Delivery: 20X TBS (Tris Buffered Saline) solution consisting of 500 mM Tris, 60 mM KCl, 2,8 M NaCl, and 1,0% Tween 20 in high purity dH₂O adjusted to pH 7,4.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together