Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Seals
Sealing Rings
Sealing rings, EPDM rubber
Sealing rings, EPDM rubber
Sealing rings, EPDM rubber
Catalog # 18-8798-01
Supplier:
Cytiva
Expand image
Sealing rings, EPDM rubber
Catalog # 18-8798-01
Supplier:
Cytiva
Specifications
Colour:
Black
Description:
Sealing ring
Material:
EPDM
Seq:
59
Environmentally Preferable:
Pk:
5
Specifications
About this item
Product Details
Documentation
Product Details
Documentation