HMDS (1,1,1,3,3,3-hexamethyldisilazane) for synthesis, Sigma-Aldrich®
: Merck
: Hexamethyldisilazane
:999-97-3
:
:(CH₃)₃SiNHSi(CH₃)₃
:MFCD00008259
:126
:–82 °C
:0.7742 g/cm³ (20 °C)
:161.39 g/mol
:6
:Ambient
:213-668-5
:3286
:3 (6.1, 8) II
:13,04708
:M-Clarity™ quality level = MQ200
:01-2119438176-38
:GHS02
:GHS06
:GHS05
- Environmentally Preferable:
- Solubility:(20 °C) decomposes
- Melting Point:-82 °C
- Molar Mass:161.39 g/mol
- Boiling Point:126 °C (1013 mbar)
- Vapor Pressure:20 hPa (20 °C)
- Flash Point:15 °C
- Explosion Limit:0.8 - 25.9 %(V)
- Density:0.78 g/cm3 (20 °C)
- Ignition Point:325 °C
- Viscosity Kinematic:0.9 mm2/s (25 °C)
Frequently Bought Together