Vinyltrimethoxysilane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Trimethoxy(vinyl)silane
CAS Number:2768-02-7
:
Formula:C₅H₁₂O₃Si
MDL Number:MFCD00008605
Boiling Pt:123 °C (760 mmHg)
Melting Pt:0 °C
Density:0.968 g/cm³
Molecular Weight:148.23 g/mol
Flash Pt:23 °C
Storage Temperature:Ambient
EINECS:220-449-8
UN:1993
ADR:3 III
:GHS02
:GHS07
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3915 to 1.3935 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together