Barium sulphate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7727-43-7
:
Formula:BaSO₄
MDL Number:MFCD00003455
Boiling Pt:1600
Melting Pt:1580 °C (dec.)
Density:4.5
Molecular Weight:233.39 g/mol
Merck Index:13,00997
Storage Temperature:Ambient
EINECS:231-784-4
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together