Magnesium oxide analytical reagent, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Magnesia usta
CAS Number:1309-48-4
:
Formula:MgO
MDL Number:MFCD00011109
Boiling Pt:3600
Melting Pt:2800
Density:3.58 g/cm³ (25 °C)
Molecular Weight:40.3 g/mol
Merck Index:13,05700
:M-Clarity™ quality level = MQ300
EINECS:215-171-9
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:2800 °C
- Molar Mass:40.3 g/mol
- Bulk Density:100 kg/m3
- Boiling Point:3600 °C (1000 hPa)
- Density:3.58 g/cm3 (25 °C)
- pH:10.3 (H2O, 20 °C) (saturated solution)
Specification Test Results
Related Information
Frequently Bought Together