9,10-Phenanthrenequinone for synthesis, Sigma-Aldrich®
: Merck
: Phenanthrene-9,10-dione
:84-11-7
:
:C₁₄H₈O₂
:MFCD00001163
:360
:203…206 °C
:1.4045 g/cm³ (20 °C)
:208.22 g/mol
:201-515-5
:3077
:9 III
:12,07355
:M-Clarity™ quality level = MQ200
:GHS07
:GHS09
:Ambient
- Environmentally Preferable:
- Solubility:7.5 mg/l (25 °C)
- Melting Point:203 - 206 °C
- Molar Mass:208.21 g/mol
- Bulk Density:350 kg/m3
- Boiling Point:360 °C
- Flash Point:245 °C
- Ignition Point:630 °C DIN 51794
Frequently Bought Together