Piperazine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-85-0
:
Formula:C₄H₁₀N₂
MDL Number:MFCD00005953
Boiling Pt:146
Melting Pt:44…46 °C
Density:1.1 g/cm³ (20 °C)
Molecular Weight:86.14 g/mol
Flash Pt:65
EINECS:203-808-3
UN:2579
ADR:8 III
Merck Index:13,07545
:M-Clarity™ quality level = MQ200
:GHS02
:GHS05
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:150 g/l (20 °C)
- Melting Point:107 - 111 °C
- Molar Mass:86.14 g/mol
- Bulk Density:400 kg/m3
- Vapor Pressure:- 10 hPa (20 °C)
- Flash Point:65 °C
- Explosion Limit:4 - 14 %(V)
- Density:1.1 g/cm3 (20 °C)
- pH:12 (150 g/l, H2O, 20 °C)
- Ignition Point:320 °C DIN 51794
- Hazard Class:8
- Hazard Class Description:Corrosive material
Specification Test Results
Related Information
Frequently Bought Together