Piperazine, anhydrous 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-85-0
:
Formula:C₄H₁₀N₂
MDL Number:MFCD00005953
Boiling Pt:146…148 °C
Melting Pt:108…113 °C
Density:1.110
Molecular Weight:86.14 g/mol
Flash Pt:88 °C (190 °F)
UN:2579
ADR:8 III
Merck Index:14,7464
EINECS:203-808-3
:GHS08
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together