4-Methoxybenzyl alcohol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Anisyl alcohol, p-Anisyl Alcohol, 4-(Hydroxymethyl)anisole, Anise alcohol, (4-Methoxyphenyl)methanol
CAS Number:105-13-5
:
Formula:C₈H₁₀O₂
MDL Number:MFCD00004653
Boiling Pt:259 °C (760 mmHg)
Melting Pt:23…25.5 °C
Density:1.1125 g/cm³
Molecular Weight:138.17 g/mol
Flash Pt:146 °C
Merck Index:13,00608
Storage Temperature:Ambient
EINECS:203-273-6
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.5440 to 1.5470 (20°C, 589 nm)
- GC:≥97.5 %
- Specific gravity:(25°C) 1.111 to 1.115
- Filt.over 0.2 micron PFTE:Filtered
- Acidity (CH3COOH):≤0.0005 meq/g
- Peroxides:≤0.0025 % (H2O2)
Specification Test Results
Frequently Bought Together