L(-)-Glutathione (oxidised form) ≥95%, high purity
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: L(-)-Glutathione, oxidized, Bi(glutathion-S-yl), GSSG, Glutathiol, Oxidized Glutathione (GSSG), Glutathione oxidized, (2S,2'S)-5,5'-(((2R,2'R)-Disulphanediylbis(1-((carboxymethyl)amino)-1-oxopropane-3,2-diyl))bis(azanediyl))bis(2-amino-5-oxopentanoic acid)
CAS Number:27025-41-8
:
Formula:C₂₀H₃₂N₆O₁₂S₂
MDL Number:MFCD00063106
Molecular Weight:612.64 g/mol
Melting Pt:185
Storage Temperature:Refrigerator
EINECS:248-170-7
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Iron:<0.002%
- Lead:<0.001%
- Loss on Drying:≤8%
- Purity:≥95%
Specification Test Results
Frequently Bought Together