Pivaloyl chloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Trimethylacetyl chloride
CAS Number:3282-30-2
:
Formula:C₅H₉ClO
MDL Number:MFCD00000709
Boiling Pt:105
Melting Pt:–56 °C
Density:0.98 g/cm³
Molecular Weight:120.58 g/mol
Flash Pt:14 °C
Storage Temperature:Ambient
EINECS:221-921-6
UN:2438
ADR:6.1 I
:GHS06
:GHS05
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light pink Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4110 to 1.4130 (20°C, 589 nm)
- GC:≥98.5 %
- Titration RCOCl:98.5 to 101.5 %
Specification Test Results
Frequently Bought Together