Ethanesulphonic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:594-45-6
:
Formula:C₂H₆O₃S
MDL Number:MFCD00007529
Boiling Pt:123 °C (1 mmHg)
Melting Pt:–17 °C
Density:1.343 g/cm³ (20 °C)
Molecular Weight:110.13 g/mol
EINECS:209-843-0
ADR:8 III
:M-Clarity™ quality level = MQ200
UN:2586
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:C2H6O3S
- Melting Point:C2H5SO3H
- Molar Mass:110.13 g/mol
- Boiling Point:136 - 140 °C (3 hPa)
- Refractive Index:1.4335 (20 °C, 589 nm)
- Density:1.34 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together