2-Mercaptoethanol 99%, pure
: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol
:60-24-2
:
:HSCH₂CH₂OH
:MFCD00004890
:157…158 °C (760 mmHg)
:–100 °C
:1.11 g/cm³
:78.14 g/mol
:73 °C
:2966
:6.1 II
:13,05894
:200-464-6
:GHS06
:GHS09
:GHS07
:GHS05
:Ambient
- Environmentally Preferable:
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4990 to 1.5020 (20°C, 589 nm)
- Water:≤0.5 %
- GC:≥98.5 %
Frequently Bought Together