1,6-Diaminohexane 99.5+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,6-Hexanediamine, Hexamethylenediamine
CAS Number:124-09-4
:
Formula:C₆H₁₆N₂
MDL Number:MFCD00008243
Boiling Pt:204…205 °C (760 mmHg)
Molecular Weight:116.21 g/mol
Melting Pt:38…41 °C
Flash Pt:81 °C
ADR:8 III
Merck Index:13,04713
EINECS:204-679-6
UN:2280
:GHS07
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline low melting mass (May discolor on storage)
- Infrared spectrum:Conforms
- Melting point:38 to 41 °C
- Water:≤0.05 % (K.F.)
- GC:≥99.50 %
- Titration with HClO4:99.5 to 100.5 %
Specification Test Results
Frequently Bought Together