Diammonium hydrogen citrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
: Citric acid diammonium salt, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Ammonium citrate dibasic
:3012-65-5
:
:HOC(CO₂H)(CH₂CO₂NH₄)₂
:MFCD00013068
:1.483 g/cm³ (20 °C)
:226.19 g/mol
:13,00513
:M-Clarity™ quality level = MQ300
:221-146-3
:GHS07
:Ambient
- Environmentally Preferable:
- Solubility:(20 °C) soluble
- Molar Mass:226.19 g/mol
- Bulk Density:400 - 600 kg/m3
- Boiling Point:(1013 hPa) (decomposition)
- Vapor Pressure:(20 °C)
- Density:1.48 g/cm3 (20 °C)
- pH:5.2 (50 g/l, H2O, 20 °C)
Frequently Bought Together