Dibutyl sebacate ≥90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sebacic acid dibutyl ester, 1,10-Decanedioic acid dibutyl ester, Dibutyl decanedioate
CAS Number:109-43-3
:
Formula:C₁₈H₃₄O₄
MDL Number:MFCD00027218
Boiling Pt:344…345 °C
Melting Pt:–11 °C
Density:0.94
Molecular Weight:314.47 g/mol
Flash Pt:178 °C (352 °F)
EINECS:203-672-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together