Ethyl butyrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Butanoic acid ethyl ester, Butyric acid ethyl ester
CAS Number:105-54-4
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00009394
Boiling Pt:119…120 °C
Melting Pt:–93 °C
Density:0.879
Molecular Weight:116.16 g/mol
Flash Pt:26 °C (79 °F)
UN:1180
ADR:3 III
Merck Index:13,03810
EINECS:203-306-4
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together