Biuret ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Carbamylurea, Allophanic acid amide
CAS Number:108-19-0
:
Formula:C₂H₅N₃O₂
MDL Number:MFCD00007946
Density:1.46
Molecular Weight:103.08 g/mol
Melting Pt:∼ 189 °C (dec.)
Merck Index:13,01305
EINECS:203-559-0
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together