Manganese(II) sulphate tetrahydrate 99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Manganese sulphate tetrahydrate, Manganous sulphate tetrahydrate
CAS Number:10101-68-5
:
Formula:MnO₄S·4H₂O
MDL Number:MFCD00149160
Density:2.11 g/cm³ (20 °C)
Molecular Weight:223.06 g/mol
Storage Temperature:Ambient
EINECS:232-089-9
UN:3077
ADR:9 III
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together