Order Entry
Italy
ContactUsLinkComponent
Barium iodide hydrate 98% for analysis
Barium iodide hydrate 98% for analysis
Catalog # ACRO208000500
CAS Number:  
undefined
Barium iodide hydrate 98% for analysis
Catalog # ACRO208000500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Titration Argentometric:
    ≥97.5 % (on dry substance)
  • Chloride (Cl):
    ≤2000 ppm
  • Chromium (Cr):
    ≤10 ppm
  • Cobalt (Co):
    ≤10 ppm
  • Copper (Cu):
    ≤12.5 ppm
  • Iron (Fe):
    ≤5 ppm
  • Lead (Pb):
    ≤100 ppm
  • Manganese (Mn):
    ≤10 ppm
  • Nickel (Ni):
    ≤10 ppm
  • Sodium (Na):
    ≤1000 ppm

Specification Test Results

About this item

Synonyms: Barium iodide dihydrate
 :
 :409.15 g/mol
EINECS:237-276-9
UN:1564
 :GHS07
CAS Number:85017-89-6
Formula:BaI₂·H₂O
MDL Number:MFCD00149155
Melting Pt:740 °C
Density:5.15
ADR:6.1 III