Chromium(III) potassium sulphate dodecahydrate 98+%
: Chromium potassium sulphate dodecahydrate, Chromium (III) potassium bis(sulphate) dodecahydrate
:7788-99-0
:
:CrK(SO₄)₂·12H₂O
:MFCD00149917
:499.4 g/mol
:89 °C
:13,02250
:233-401-6
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Violet to dark purple Crystalline powder or crystals
- Titration Iodometric:98.0 to 102.0 %
- Chloride (Cl):≤20 ppm
- Aluminium (Al):≤200 ppm
- Ammonium (NH4):≤100 ppm
- Iron (Fe):≤100 ppm
- Insoluble matter:≤0.01 % (in water)
Frequently Bought Together