Oxalic acid dihydrate, high purity
: Ethanedioic acid dihydrate
:6153-56-6
:
:HO₂CCO₂H·2H₂O
:MFCD00149102
:149…160 °C (1013 hPa)
:101
:1.65 g/cm³ (20 °C)
:126.07 g/mol
:Ambient
:205-634-3
:8 III
:13,06980
:3261
:01-2119534576-33
:GHS07
- Environmentally Preferable:
- Ash:<0.2%
- Chloride:<0.005%
- Insolubles:<0.05%
- Iron:<0.005%
- Purity:>99%
Frequently Bought Together