Oxalic acid dihydrate 99.5-102.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ethanedioic acid dihydrate
CAS Number:6153-56-6
:
Formula:HO₂CCO₂H·2H₂O
MDL Number:MFCD00149102
Boiling Pt:149…160 °C (1013 hPa)
Melting Pt:101…105 °C
Density:1.65
Molecular Weight:126.07 g/mol
Storage Temperature:Ambient
EINECS:205-634-3
ADR:8 III
Merck Index:14,6911
UN:3261
REACH:01-2119534576-33
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together