N-(1-Naphthyl)ethylenediamine dihydrochloride ≥95%, GPR RECTAPUR®
Catalog # 293044M
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:10 g
- Seq:3236
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (calculated on dried substance):Min. 95 %
Specification Test Results
About this item
Synonyms: Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine
:
:259.18 g/mol
EINECS:215-981-2
:GHS07
Storage Temperature:Ambient
CAS Number:1465-25-4
Formula:C₁₂H₁₆Cl₂N₂
MDL Number:MFCD00012556
Melting Pt:188…190 °C
Merck Index:12,06496