Order Entry
Italy
ContactUsLinkComponent
 
Methanesulphonic acid 70% ≥70% in aqueous solution for synthesis, Sigma-Aldrich®
 :  Merck
 
undefined
Methanesulphonic acid 70% ≥70% in aqueous solution for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Environmentally Preferable:

 

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -60 °C
  • Density:
    1.35 g/cm3 (20 °C)

 

Frequently Bought Together