Order Entry
Italy
ContactUsLinkComponent
DL-Methionine
DL-Methionine
Catalog # EHERC15021000
Supplier:  EHRENSTORFER
CAS Number:  
DL-Methionine
Catalog # EHERC15021000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 2-Amino-4-(methylthio)butanoic acid, (±)-2-Amino-4-(methylthio)butyric acid, Methioninum racemicum, Methionine racematic, H-DL-Met-OH, (±)-Methionine, (±)-2-Amino-4-(methylthio)butanoic acid
 :
 :149.21 g/mol
Melting Pt:∼136 °C
Density:1.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-432-1
CAS Number:59-51-8
Formula:C₅H₁₁NO₂S
MDL Number:MFCD00063096
Merck Index:13,06004