Order Entry
Italy
ContactUsLinkComponent
Nickel chromium alloy (80:20 w%) ≥99.95% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Nickel chromium alloy (80:20 w%) ≥99.95% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Catalog # 42366.18
CAS Number:  
Nickel chromium alloy (80:20 w%) ≥99.95% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Catalog # 42366.18
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368
  • Environmentally Preferable:

Specifications

About this item

 :
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:11106-97-1
MDL Number:MFCD00673110