Order Entry
Italy
ContactUsLinkComponent
Acrylamide : Bisacrylamide (37.5:1) 40% in aqueous solution
Acrylamide : Bisacrylamide (37.5:1) 40% in aqueous solution
Catalog # A4989.0500
Supplier:  PanReac AppliChem
CAS Number:  
Acrylamide : Bisacrylamide (37.5:1) 40% in aqueous solution
Catalog # A4989.0500
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Acrylamide- N,N'-Methylenebisacrylamide, Acrylamide-Bis
 :
UN:3426
 :GHS06
 :GHS08
Storage Temperature:Ambient
 :79-06-1
MDL Number:MFCD00080848
ADR:6.1 III