D-(+)-Maltose monohydrate ≥85%, Reagent Grade
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
CAS Number:6363-53-7
:
Formula:C₁₂H₂₂O₁₁·1H₂O
MDL Number:MFCD00149343
Molecular Weight:360.32 g/mol
Melting Pt:160…165 °C
Merck Index:13,05736
Storage Temperature:Ambient
EINECS:200-716-5
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Loss on Drying:< 7%
- Purity:> 85 %
- Residue after Ignition:< 0.1 %
Specification Test Results
Frequently Bought Together