Ethyl oleate mixture of homologeous fatty acid esters 98%
: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester
:111-62-6
:85049-36-1
:C₂₀H₃₈O₂
:MFCD00009579
:190…210 °C (7 mmHg)
:-32
:0.86 g/cm³
:310.52 g/mol
:160 °C
:12,06965
:Ambient
- Environmentally Preferable:
- Appearance:Clear viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4490 to 1.4530 (20°C, 589 nm)
- Assay:≥97.5 % (ester)
- Water:≤1 % (K.F.)
- Density:(20°C) 0.869 to 0.874
- GC:≥60 % (oleic acid)
- Color scale:≤70 APHA
- Stabilizer:80 to 120 ppm BHA content
- Acid value:≤0.5 mg KOH/g
- Saponification value:177 to 188 mg KOH/g
Frequently Bought Together