Anti-Tubulin Beta-3 Mouse Monoclonal Antibody
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Host: Mouse, Isotype: IgG1
- For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Recognizes human betaIII-tubulin located in neurons.
Type: Primary
Antigen: TUBULIN BETA-3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ENZOALX804405C100
- Antigen symbol:
- TUBB3
- Antigen name:
- Tubulin Beta 3
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Host:
- Mouse
- Isotype:
- IgG1
- Western blot:
- Yes
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Flow cytometry:
- Yes
- Form:
- Liquid. In PBS, pH 7,4, containing 15 mM sodium azide
- Purification:
- ≥95% (SDS-PAGE)
- Concentration:
- 1 mg/ml
Specifications
Frequently Bought Together