Anti-DHFR/Dihydrofolate Reductase Polyclonal Antibody
Catalog # BSBTPB9175
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:DHFR/Dihydrofolate Reductase
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for Dihydrofolate Reductase/DHFR detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. Dihydrofolate Reductase/DHFR information: Molecular Weight: 21453 MW; Tissue Specificity: Widely expressed in fetal and adult tissues, including throughout the fetal and adult brains and whole blood. Expression is higher in the adult brain than in the fetal brain.
Type: Primary
Antigen: DHFR/Dihydrofolate Reductase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat