Anti-CCL17/TARC Polyclonal Antibody
Catalog # BSBTPB9031
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:CCL17/TARC
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for TARC/CCL17 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: ELISA. Reactive species: Mouse. TARC/CCL17 information: Molecular Weight: 10467 MW; Subcellular Localization: Secreted .
Type: Primary
Antigen: CCL17/TARC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse