Brimonidine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: UK 14,304, 5-Bromo-N-(2-imidazolin-2-yl)-6-quinoxalinamine, 5-Bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine
CAS Number:59803-98-4
:
Formula:C₁₁H₁₀BrN₅
MDL Number:MFCD00153878
Molecular Weight:292.14 g/mol
Melting Pt:235…236 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1 III
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Residue after ignition:≤0.3 %
Specification Test Results
Frequently Bought Together