Order Entry
Italy
ContactUsLinkComponent
Suxamethonium chloride 96%
Suxamethonium chloride 96%
Certificates
undefined
Suxamethonium chloride 96%
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    160 to 166 °C
  • Titration Argentometric:
    ≥95 % (on dry substance)
  • Water:
    ≤10 %

Specification Test Results

Related Information

Frequently Bought Together