Anti-EGF Rabbit Polyclonal Antibody (Biotin)
PRSIXP-5119BT
- Antigen name:Epidermal growth factor
- Antigen symbol:EGF
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:kinesin-related motor protein Eg5|thyroid receptor-interacting protein 5|TRIP5|TR-interacting protein 5|kinesin-like spindle protein HKSP|HKSP|kinesin-like 1|thyroid receptor interacting protein 5|kinesin-like protein 1|KNSL1|TRIP-5|kinesin-like protein KIF11
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hEGF (human Epidermal Growth Factor).
- Size:50 µg
- Pk:50 µG
Anti-hEGF specific antibody was purified by affinity chromatography and then biotinylated.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human