Specifications
- Antigen name:Cell Division Cycle 25A
- Antigen symbol:CDC25A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Dual specificity phosphatase Cdc25A|CDC25A|M-phase inducer phosphatase 1
- Concentration:1 mg/mL
- Immunogen:cdc25A (Phospho-Ser76) antibody was raised against a peptide sequence around phosphorylation site of serine 76 (M-G-S (p) -S-E) derived from Human cdc25A.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Tyrosine protein phosphatase which functions as a dosage-dependent inducer of mitotic progression. Directly dephosphorylates CDC2 and stimulates its kinase activity. Also dephosphorylates CDK2 in complex with cyclin E, in vitro.
Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100
Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CDC25A
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: