Anti-ADRB3 Chicken Polyclonal Antibody
Catalog # PRSI54-197
Supplier: ProSci Inc.
Specifications
- Antigen name:Adrenoceptor Beta 3
- Antigen symbol:ADRB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Adrb-3|FLJ99960|BETA3AR
- Immunogen:MouseADRB3 peptide AA386-400
- Size:100 µl
- Pk:100 µl
Specifications
About this item
ADRB3 antibody can be used at 1:2000-1:5000 dilutions for Western Blot analysis and for other immuno-assays. Customers need to optimize the dilution when using the antibody in IHC or ICC.
Affinity Purified
PBS Buffer containing 0.02% sodium azide.
2-8°C for one week and -20°C as an undiluted liquid for up to two years.
Type:
Antigen: ADRB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Mouse, Rat