Specifications
- Antigen name:RAD52 homolog
- Antigen symbol:RAD52
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:RAD52 homolog S. cerevisiae|DNA repair protein RAD52 homolog|Recombination protein RAD52|RAD52 homolog|Rhabdomyosarcoma antigen MU RMS 40.23.|RAD 52|DNA repair protein RAD52
- Immunogen:RAD52 antibody was raised against amino acids 122 - 135 of RAD52 (Human).
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
The protein encoded by this gene shares similarity with Saccharomyces cerevisiae Rad52, a protein important for DNA double-strand break repair and homologous recombination. This gene product was shown to bind single-stranded DNA ends, and mediate the DNA-DNA interaction necessary for the annealing of complementary DNA strands. It was also found to interact with DNA recombination protein RAD51, which suggested its role in RAD51 related DNA recombination and repair. Four alternatively spliced transcript variants encoding different isoforms have been reported for this gene. The alpha variant encodes the longest isoform that contains an identical N-terminus, but a distinct C-terminus, as compared to other isoforms (beta, delta, and gamma).
0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.
Store RAD52 antibody at -20 °C prior to opening. Dilute only prior to immediate use. For extended storage aliquot contents and freeze at -20 °C or below. As with all antibodies avoid freeze/thaw cycles.
Type:
Antigen: RAD52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human