Order Entry
Italy
ContactUsLinkComponent
 
Anti-MPZ Goat Polyclonal Antibody
  PRSI45-925
undefined
Anti-MPZ Goat Polyclonal Antibody
  PRSI45-925

 

  • Antigen name:
    Myelin Protein Zero
  • Antigen symbol:
    MPZ
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    P0|myelin protein zero (Charcot-Marie-Tooth neuropathy 1B) |CHM|CMT1B|CMT4E|MPZ|HMSNIB|CMT2I|myelin protein zero|MPP|CMTDI3|CMT2J|DSS|CMT1
  • Concentration:
    500 ug/mL
  • Immunogen:
    Myelin Protein Zero antibody was raised against a 12 amino acid synthetic peptide near the internal region (near the C-Terminus) of Myelin Protein Zero.
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Myelin Protein Zero antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Myelin Protein Zero antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MPZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat