Anti-ELF5 Goat Polyclonal Antibody
PRSI45-528
- Antigen name:E74-like Factor 5 (ets Domain transcription Factor)
- Antigen symbol:ELF5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:ESE-2|ETS-related transcription factor Elf-5|Epithelium-restricted ESE-1-related Ets factor|Epithelium-specific Ets transcription factor 2|ESE2|ELF5|E74-like factor 5
- Concentration:500 ug/mL
- Immunogen:ELF5 antibody was raised against a 12 amino acid synthetic peptide near the C-Terminus of ELF5.
- Size:100 µg
- Pk:100 µG
ELF5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ELF5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ELF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human