Order Entry
Italy
ContactUsLinkComponent
Anti-WIZ Goat Polyclonal Antibody
Anti-WIZ Goat Polyclonal Antibody
Catalog # PRSI42-065
Supplier:  ProSci Inc.
undefined
Anti-WIZ Goat Polyclonal Antibody
Catalog # PRSI42-065
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Wdely interspaced zinc Finger motifs
  • Antigen symbol:
    WIZ
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    ZNF803|WIZ|Protein Wiz|Widely-interspaced zinc finger-containing protein
  • Concentration:
    500 ug/mL
  • Immunogen:
    WIZ antibody was raised against a 15 amino acid synthetic peptide near the internal region of WIZ.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

WIZ antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

WIZ antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: WIZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human