Anti-LARP1 Rabbit Polyclonal Antibody
Catalog # PRSI29-485
Supplier: ProSci Inc.
Specifications
- Antigen name:La ribonucleoprotein Domain Family member 1
- Antigen symbol:LARP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:LARP
- Immunogen:A synthetic peptide corresponding a region of human LARP1.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
LARP1 belongs to the LARP family and it contains 1 HTH La-type RNA-binding domain. The exact function of LARP1 remains unknown.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LARP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human