Specifications
- Antigen name:Triosephosphate Isomerase 1
- Antigen symbol:TPI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:TPI1|triosephosphate isomerase 1|HEL-S-49|TPI|TIM
- Immunogen:A synthetic peptide corresponding a region of human TPI1.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
TPI1 belongs to the triosephosphate isomerase family. Defects in TPI1 are the cause of triosephosphate isomerase deficiency (TPI deficiency). TPI deficiency is an autosomal recessive disorder. It is the most severe clinical disorder of glycolysis. It is associated with neonatal jaundice, chronic hemolytic anemia, progressive neuromuscular dysfunction, cardiomyopathy and increased susceptibility to infection.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TPI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human