Anti-GALNT5 Rabbit Polyclonal Antibody
Catalog # PRSI26-182
Supplier: ProSci Inc.
Specifications
- Antigen name:Polypeptide N-Acetylgalactosaminyltransferase 5
- Antigen symbol:GALNT5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:GALNAC-T5|GALNACT5
- Immunogen:A synthetic peptide corresponding a region of human GALNT5.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
GALNT5 can catalyze the initial reaction in O-linked oligosaccharide biosynthesis,the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor. GALNT5 has activity toward EA2 peptide substrate, but it has a weak activity toward Muc2 or Muc1b substrates.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GALNT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat