- Antibody type:Primary
- Antigen name:Activating Transcription Factor 2
- Antigen symbol:ATF2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:1386
- Antigen synonyms:Cyclic AMP-dependent transcription factor ATF-2|HB16|cAMP-dependent transcription factor ATF-2|CREBP1|Histone acetyltransferase ATF2|ATF-2|CREB2|cAMP-responsive element-binding protein 2|CREB-2|cAMP response element-binding protein CRE-BP1|Activating transcription factor 2|Cyclic AMP-responsive element-binding protein 2
- Modification:phospho-Thr55
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds to the cAMP-responsive element (CRE), an octameric palindrome. It forms a homodimer or a heterodimer with c-Jun and stimulates CRE-dependent transcription. This protein is also a histone acetyltransferase (HAT) that specifically acetylates histones H2B and H4 in vitro; thus it may represent a class of sequence-specific factors that activate transcription by direct effects on chromatin components. Several alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Jan 2012].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ATF2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat