Anti-HMGN1 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-5384R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:High-mobility Group Nucleosome Binding Domain 1
- Antigen symbol:HMGN1
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:3150
- Antigen synonyms:MGC117425|FLJ31471|MGC104230|HMG14|FLJ27265
- Modification:phospho-Ser21/Ser25
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Chromosomal protein HMG14 binds to the inner side of the nucleosomal DNA, thus altering the interaction between the DNA and the histone octamer and is involved in the process that maintains transcribable genes in an appropriate chromatin conformation. HMG14 has been implicated as a contributing factor in the etiology of Down syndrome.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HMGN1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat