Anti-CERS2 Rabbit Polyclonal Antibody (Cy5®)
BOSSBS-5077R-CY5
: Bioss
- Antibody type:Primary
- Antigen name:Ceramide synthase 2
- Antigen symbol:CERS2
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:29956
- Antigen synonyms:FLJ10243|SP260|CerS2|MGC987|LAG1 homolog ceramide Synthase 2|L3|TMSG1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:230-261/380
- Purification:Purified by Protein A
- Pk:100 µl
LASS2 (LAG1 homolog, ceramide synthase 2) has sequence similarity to yeast longevity assurance gene 1. It is thought to be involved in sphingolipid synthesis. Expression of LASS2 is transiently increased during the period of active myelination and is specifically localized to white matter tracts of the brain, including the Schwann cells of sciatic nerves suggesting that LASS2 is important for the synthesis of dihydroceramide used for synthesis of myelin sphingolipids. Expression of LASS2 in hepatocellular carcinoma cell lines suppresses the growth of cancer cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CERS2
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: