- Antibody type:Primary
- Antigen name:SHC (Src Homology 2 Domain Containing) Transforming Protein 2
- Antigen symbol:SHC2
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Src homology 2 domain-containing-transforming protein C2|Protein Sck|SHCB|SHC2|SHC-transforming protein 2|SHC-transforming protein B|SH2 domain protein C2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Src homology 2 (SH2) domains bind specifically to tyrosine-phosphorylated proteins that temporally participate in signal transduction events (1). Shc-like protein (Sck) is a neuronal adaptor protein that contains an N-terminal PTB (phosphotyrosine binding) domain, a collagen homology (CH) domain, and a conserved C-terminal SH2 domain (2,3). Human Sck transcripts are present at high levels in liver, pancreas, prostate and ovary (4,5). In vascular endothelial cells, Sck participates in VEGF-induced signal transduction (6). Treatment of human umbilical vein endothelial (HUVEC) cells with VEGF induces recruitment of Sck to tyrosine-1175 of the kinase insert domain-containing receptor (KDR) and enhances Sck tyrosine phosphorylation (7,8).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SHC2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat